Learn More
Selleck Chemical LLC 5-Ph-IAA E8177-5MG
SDP
Catalog No. 5000043720
Encompass_Preferred
The 5-Ph-IAA is a bumped-IAA analogue that establishes the auxin-inducible degron 2 (AID2) system with the OsTIR1(F74G) mutant enabling precise rapid degradation with 670-fold lower ligand concentration It induces rapid and efficient depletion of mAID-fused proteins to study protein function in living cells exhibiting significant tumor suppression properties
Catalog No. 50-000-43720 Supplier Selleck Chemical LLC Supplier No. E81775MG
The 5-Ph-IAA is a bumped-IAA analogue that establishes the auxin-inducible degron 2 (AID2) system with the OsTIR1(F74G) mutant enabling precise rapid degradation with 670-fold lower ligand concentration It induces rapid and efficient depletion of mAID-fused proteins to study protein function in living cells exhibiting significant tumor suppression properties