Learn More
TARGETMOL CHEMICALS INC 5-PH-IAA 25MG
SDP
Catalog No. 502579039
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 5-Ph-IAA is a derivative of Indole-3-acetic acid (IAA) which is a plant hormone and acts as an enzyme or prodrug combination for cancer gene therapy. purity: 99%
Catalog No. 50-257-9039 Supplier TARGETMOL CHEMICALS INC Supplier No. T888525MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 5-Ph-IAA is a derivative of Indole-3-acetic acid (IAA) which is a plant hormone and acts as an enzyme or prodrug combination for cancer gene therapy. purity: 99%