Learn More
Medchemexpress LLC 5-ph-IAA-AM | 2990123-10-7 | 99.0% | C19H17NO4 | 100 MG
SDP
Catalog No. 5000354571
Encompass_Preferred
5-Ph-IAA-AM is an eggshell-permeable analog that contains an acetoxymethyl (AM) group. It provides enhanced protein degradation in laid embryos and can be utilized to reveal the roles of proteins in *C. elegans*, particularly those involved in embryogenesis and development, through temporally controlled protein degradation.
  • Eggshell-permeable analog
  • Contains an acetoxymethyl (AM) group
  • Provides enhanced protein degradation in laid embryos
  • Useful for understanding protein roles in *C. elegans*
  • Supports research into embryogenesis and development
  • Enables temporally controlled protein degradation
Catalog No. 50-003-54571 Supplier Medchemexpress LLC Supplier No. HY141894100MG
5-Ph-IAA-AM is an eggshell-permeable analog that contains an acetoxymethyl (AM) group. It provides enhanced protein degradation in laid embryos and can be utilized to reveal the roles of proteins in *C. elegans*, particularly those involved in embryogenesis and development, through temporally controlled protein degradation.
  • Eggshell-permeable analog
  • Contains an acetoxymethyl (AM) group
  • Provides enhanced protein degradation in laid embryos
  • Useful for understanding protein roles in *C. elegans*
  • Supports research into embryogenesis and development
  • Enables temporally controlled protein degradation