Learn More
Medchemexpress LLC 5-Ph-IAA-AM | 2990123-10-7 | MFCD30097746 | 99.04% | C19H17NO4 | 50 MG
SDP
Catalog No. 5000354531
Encompass_Preferred
5-Ph-IAA-AM is an eggshell-permeable analog of 5-Ph-IAA containing an acetoxymethyl (AM) group. It provides enhanced protein degradation in laid embryos and can be used to uncover the roles of proteins in *C. elegans*, particularly those involved in embryogenesis and development, through temporally controlled protein degradation.
  • Induced better protein degradation in *C. elegans* embryos compared to 5-Ph-IAA.
  • Embryos injected with 5-Ph-IAA-AM failed to form ventral furrows or undergo other gastrulation processes, consistent with the loss of myosin function in early *Drosophila* embryos.
Catalog No. 50-003-54531 Supplier Medchemexpress LLC Supplier No. HY14189450MG
5-Ph-IAA-AM is an eggshell-permeable analog of 5-Ph-IAA containing an acetoxymethyl (AM) group. It provides enhanced protein degradation in laid embryos and can be used to uncover the roles of proteins in *C. elegans*, particularly those involved in embryogenesis and development, through temporally controlled protein degradation.
  • Induced better protein degradation in *C. elegans* embryos compared to 5-Ph-IAA.
  • Embryos injected with 5-Ph-IAA-AM failed to form ventral furrows or undergo other gastrulation processes, consistent with the loss of myosin function in early *Drosophila* embryos.