Learn More
Medchemexpress LLC HY-141894 5mg , 5-Ph-IAA-AM CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1418945MG
Encompass_Preferred
HY-141894 5mg 5-Ph-IAA-AM CAS: 5-Ph-IAA-AM is an eggshell-permeable 5-Ph-IAA analog. 5-Ph-IAA-AM affords an enhanced protein degradation in laid embryos. 5-Ph-IAA-AM can be used to disclosure the roles of proteins in C. elegans, in particular those that are involved in embryogenesis and development, through temporally controlled protein degradation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1846