Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-phenyl-1-pentyne | 1823-14-9 | MFCD00039813 | 98.0% | 144.21 g/mol | C11H12 | 1 G
SDP

Catalog No. 5000283373
Encompass_Preferred

5-Phenyl-1-pentyne is an aromatic alkyne reagent used as a synthetic intermediate and research standard. It is provided at high purity for organic synthesis and analytical applications, and is suitable for small-scale laboratory work.

  • Has 98.0% purity.
  • Chemical formula C11H12.
  • Molecular weight 144.21 g/mol.
  • CAS number 1823-14-9.
  • Available in small pack sizes suitable for laboratory use.
  • Suitable for use as a synthetic intermediate and reference reagent.

Catalog No. 50-002-83373 Supplier Medchemexpress LLC Supplier No. HYW1309011G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Phenyl-1-pentyne is an aromatic alkyne reagent used as a synthetic intermediate and research standard. It is provided at high purity for organic synthesis and analytical applications, and is suitable for small-scale laboratory work.

  • Has 98.0% purity.
  • Chemical formula C11H12.
  • Molecular weight 144.21 g/mol.
  • CAS number 1823-14-9.
  • Available in small pack sizes suitable for laboratory use.
  • Suitable for use as a synthetic intermediate and reference reagent.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.