Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Phenyl-1-pentyne, 98+%

Catalog No. aal1160806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1160806 5 g
AAL1160814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1160806 Supplier Thermo Scientific Chemicals Supplier No. L1160806
Only null left
Add to Cart

CAS: 1823-14-9 | C11H12 | 144.22 g/mol

5-Phenyl-1-pentyne is also used as an intermediate in organic synthesis and pharmaceuticals. It is also used to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Phenyl-1-pentyne is also used as an intermediate in organic synthesis and pharmaceuticals. It is also used to make other chemicals.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1823-14-9
Molecular Formula C11H12
Molecular Weight (g/mol) 144.22
MDL Number MFCD00039813
InChI Key KOSORCNALVBYBP-UHFFFAOYSA-N
Synonym 5-phenyl-1-pentyne, pent-4-yn-1-ylbenzene, benzene, 4-pentynyl, 5-phenylpentyne, unii-b4qv76z86q, pent-4-ynyl-benzene, ccris 7101, pent-4-yn-1-yl benzene, 5-phenyl pentyne, 4-pentynylbenzene #
PubChem CID 74573
IUPAC Name pent-4-ynylbenzene
SMILES C#CCCCC1=CC=CC=C1

Specifications

Density 0.95
Boiling Point 94°C (50 mmHg)
Flash Point 85°C (185°F)
Refractive Index 1.5165
Linear Formula C6H5(CH2)3C≡CH
Quantity 5 g
Beilstein 2074729
Formula Weight 144.22
Percent Purity ≥98%
Chemical Name or Material 5-Phenyl-1-pentyne
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 217-352-8
RTECSNumber DA6820000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.