Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-phenyl-1-pentyne (pent-4-ynylbenzene) | 1823-14-9 | MFCD00039813 | 98.0% | 144.21 g·mol⁻¹ | C11H12 | 500 MG
SDP

Catalog No. 5000283409
Encompass_Preferred

5-Phenyl-1-pentyne is a terminal alkyne used as a building block and intermediate in organic synthesis and medicinal chemistry research. It is supplied as a high-purity small-molecule suitable for laboratory applications.

  • High purity: 98.0%.
  • CAS number: 1823-14-9.
  • Chemical formula C11H12; molecular weight 144.21 g·mol⁻¹.
  • Provided in milligram and gram pack sizes for research use.
  • SMILES identifier available for structure confirmation.

Catalog No. 50-002-83409 Supplier Medchemexpress LLC Supplier No. HYW130901500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Phenyl-1-pentyne is a terminal alkyne used as a building block and intermediate in organic synthesis and medicinal chemistry research. It is supplied as a high-purity small-molecule suitable for laboratory applications.

  • High purity: 98.0%.
  • CAS number: 1823-14-9.
  • Chemical formula C11H12; molecular weight 144.21 g·mol⁻¹.
  • Provided in milligram and gram pack sizes for research use.
  • SMILES identifier available for structure confirmation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.