Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Phenyl-2-furancarboxylic Acid 96.0+%, TCI America™
SDP

Catalog No. P13941G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P13941G Supplier TCI America Supplier No. P13941G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 52938-97-3
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.18
MDL Number MFCD00456304
InChI Key GUOMINFEASCICM-UHFFFAOYSA-N
Synonym 5-Phenyl-2-furoic Acid
PubChem CID 736255
IUPAC Name 5-phenylfuran-2-carboxylic acid
SMILES OC(=O)C1=CC=C(O1)C1=CC=CC=C1

Specifications

Melting Point 151°C
Color White-Yellow
Quantity 1 g
Formula Weight 188.18
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Phenyl-2-furancarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.