Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-phenylimidazolidine-2,4-dione | 89-24-7 | MFCD00037881 | ≥97.0% | 176.18 g/mol | C9H8N2O2 | 100MG
SDP

Catalog No. 5000238285
Encompass_Preferred

5-Phenylhydantoin (CAS 89-24-7) is a small-molecule research compound described as having antiepileptic activity and reported hypolipidemic properties. It is supplied for research use only and is typically provided as a solid with high purity suitable for biochemical and pharmacological studies.

  • High purity: ≥97.0%.
  • Molecular formula: C9H8N2O2.
  • Molecular weight: 176.18 g/mol.
  • Physical form: solid powder or crystals.
  • Research applications: anticonvulsant and lipid metabolism studies.
  • Available in small-scale packs for lab use.
  • Not for human consumption; research use only.

Catalog No. 50-002-38285 Supplier Medchemexpress LLC Supplier No. HY125780100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Phenylhydantoin (CAS 89-24-7) is a small-molecule research compound described as having antiepileptic activity and reported hypolipidemic properties. It is supplied for research use only and is typically provided as a solid with high purity suitable for biochemical and pharmacological studies.

  • High purity: ≥97.0%.
  • Molecular formula: C9H8N2O2.
  • Molecular weight: 176.18 g/mol.
  • Physical form: solid powder or crystals.
  • Research applications: anticonvulsant and lipid metabolism studies.
  • Available in small-scale packs for lab use.
  • Not for human consumption; research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.