Learn More
Medchemexpress LLC 5-[(phenylmethoxy)methyl]-7-(1-pyrrolidinylmethyl)-8-quinolinol | 383392-66-3 | 98.9% | 348.44 | C22H24N2O2 | 5 MG
SDP
Catalog No. 5000264388
Encompass_Preferred
UC-112 is a small-molecule inhibitor of inhibitor of apoptosis proteins (IAP) developed for cellular studies of apoptosis and anticancer activity. It produces cytotoxic and antiproliferative effects across multiple human cancer cell lines and is supplied as a solid or as a DMSO solution for research applications.
  • Potent IAP inhibitor with reported cell-assay IC50 = 0.7-3.4 μM.
  • Demonstrated activity in PC-3, A-375, M14, and DU-145 cell lines.
  • High purity (98.92%) suitable for in vitro studies.
  • Molecular formula C22H24N2O2; molecular weight 348.44.
  • Available as milligram quantities and as a 10 mM solution in DMSO.
  • Intended for research use in apoptosis and anticancer mechanism studies.
Catalog No. 50-002-64388 Supplier Medchemexpress LLC Supplier No. HY128425MG
UC-112 is a small-molecule inhibitor of inhibitor of apoptosis proteins (IAP) developed for cellular studies of apoptosis and anticancer activity. It produces cytotoxic and antiproliferative effects across multiple human cancer cell lines and is supplied as a solid or as a DMSO solution for research applications.
  • Potent IAP inhibitor with reported cell-assay IC50 = 0.7-3.4 μM.
  • Demonstrated activity in PC-3, A-375, M14, and DU-145 cell lines.
  • High purity (98.92%) suitable for in vitro studies.
  • Molecular formula C22H24N2O2; molecular weight 348.44.
  • Available as milligram quantities and as a 10 mM solution in DMSO.
  • Intended for research use in apoptosis and anticancer mechanism studies.