Learn More
Medchemexpress LLC 5-phenylvaleric acid | 2270-20-4 | MFCD00004416 | 98.1% | 178.23 g/mol | C11H14O2 | 25 MG
SDP
Catalog No. 5000302977
Encompass_Preferred
5-Phenylvaleric acid (5-phenylpentanoic acid) is a small-molecule analytical standard used in chemical research and metabolite analysis. Supplied as an off-white to light brown solid, it has molecular formula C11H14O2, molecular weight 178.23 g/mol, and a reported purity of approximately 98%, with storage recommendations for long-term stability.
  • Analytical standard for research and metabolite profiling.
  • Reported purity 98.14%, appropriate for analytical workflows.
  • Off-white to light brown solid, convenient for solid handling and storage.
  • Stable under recommended storage: powder at -20°C for up to 3 years.
  • Provides well-characterized molecular data: formula C11H14O2, MW 178.23 g/mol.
Catalog No. 50-003-02977 Supplier Medchemexpress LLC Supplier No. 25MGHYW032915
5-Phenylvaleric acid (5-phenylpentanoic acid) is a small-molecule analytical standard used in chemical research and metabolite analysis. Supplied as an off-white to light brown solid, it has molecular formula C11H14O2, molecular weight 178.23 g/mol, and a reported purity of approximately 98%, with storage recommendations for long-term stability.
  • Analytical standard for research and metabolite profiling.
  • Reported purity 98.14%, appropriate for analytical workflows.
  • Off-white to light brown solid, convenient for solid handling and storage.
  • Stable under recommended storage: powder at -20°C for up to 3 years.
  • Provides well-characterized molecular data: formula C11H14O2, MW 178.23 g/mol.