Learn More
Medchemexpress LLC 5-phenylvaleric acid | 2270-20-4 | MFCD00004416 | 98.1% | 178.23 g/mol | C11H14O2 | 5 MG
SDP
Catalog No. 5000300203
Encompass_Preferred
5-Phenylvaleric acid (5-phenylpentanoic acid, CAS 2270-20-4) is a phenyl-substituted valeric acid found as an endogenous metabolite produced by gut microbiota during flavan-3-ol metabolism. It is used as an analytical standard and as a chemical building block in synthesis, and is commonly employed in biomarker and analytical studies. Molecular formula: C11H14O2; molecular weight: 178.23 g/mol.
Catalog No. 50-003-00203 Supplier Medchemexpress LLC Supplier No. HYW0329155MG
5-Phenylvaleric acid (5-phenylpentanoic acid, CAS 2270-20-4) is a phenyl-substituted valeric acid found as an endogenous metabolite produced by gut microbiota during flavan-3-ol metabolism. It is used as an analytical standard and as a chemical building block in synthesis, and is commonly employed in biomarker and analytical studies. Molecular formula: C11H14O2; molecular weight: 178.23 g/mol.