Learn More
Medchemexpress LLC 5-phenylvaleric acid | 2270-20-4 | MFCD00009109 | 98.1% | 178.23 g·mol⁻¹ | C11H14O2 | 50 MG
SDP
Catalog No. 5000299372
Encompass_Preferred
5-Phenylvaleric acid is a small-molecule analytical standard and endogenous metabolite produced by gut microbiota during flavan-3-ol metabolism. It is supplied as a solid for research and analytical applications and can be used as a synthetic intermediate or reference standard in quantification studies.
  • High purity (≈98.1%) suitable for analytical use.
  • Molecular formula C11H14O2; molecular weight 178.23 g·mol⁻¹.
  • Supplied in multiple small package sizes for analytical workflows.
  • Stable as powder at -20°C for long-term storage; in solution store at -80°C for short-term.
  • Relevant as a biomarker of flavan-3-ol metabolism in biological studies.
  • Usable as a synthetic intermediate in organic synthesis.
Catalog No. 50-002-99372 Supplier Medchemexpress LLC Supplier No. HYW03291550MG
5-Phenylvaleric acid is a small-molecule analytical standard and endogenous metabolite produced by gut microbiota during flavan-3-ol metabolism. It is supplied as a solid for research and analytical applications and can be used as a synthetic intermediate or reference standard in quantification studies.
  • High purity (≈98.1%) suitable for analytical use.
  • Molecular formula C11H14O2; molecular weight 178.23 g·mol⁻¹.
  • Supplied in multiple small package sizes for analytical workflows.
  • Stable as powder at -20°C for long-term storage; in solution store at -80°C for short-term.
  • Relevant as a biomarker of flavan-3-ol metabolism in biological studies.
  • Usable as a synthetic intermediate in organic synthesis.