Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Phenylvaleric Acid 99.0+%, TCI America™
SDP

Catalog No. P12845G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12845G 5 g
P128425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12845G Supplier TCI America Supplier No. P12845G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2270-20-4
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00004416
InChI Key BYHDDXPKOZIZRV-UHFFFAOYSA-N
Synonym 5-phenylvaleric acid, benzenepentanoic acid, phenylvaleric acid, phenylpentanoic acid, 5-phenylvalerate, valeric acid, 5-phenyl, 5-phenyl-pentanoic acid, 5-phenylvalericacid, unii-xyj5u8lcq8, 5-phenzylvaleric acid
PubChem CID 16757
ChEBI CHEBI:40131
IUPAC Name 5-phenylpentanoic acid
SMILES C1=CC=C(C=C1)CCCCC(=O)O

Specifications

Melting Point 60°C
Color White
Boiling Point 164°C
Quantity 5 g
Formula Weight 178.23
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Phenylvaleric Acid
RTECSNumber YV7816000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.