Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5'-Phosphoguanylyl-(3',5')-guanosine | 33008-99-0 | MFCD00064360 | 95.8% | 708.43 g/mol | C20H26N10O15P2 | 5 MG
SDP

Catalog No. 5000222966
Encompass_Preferred

5'-Phosphoguanylyl-(3',5')-guanosine (pGpG) is a nucleotide intermediate produced during enzymatic degradation of cyclic di-guanylate (c-di-GMP). It is supplied as a purified research reagent for biochemical and microbiology studies involving nucleotide signaling and enzymology.

  • Purity 95.8% suitable for analytical and biochemical assays.
  • Molecular weight 708.43 g/mol.
  • Chemical formula C20H26N10O15P2.
  • Available in milligram scales (5 mg, 10 mg, 25 mg, 50 mg, and larger sizes).
  • Intended for biochemical assays and studies of nucleotide signaling and c-di-GMP metabolism.
  • Provided with characterization data to support research use.

Catalog No. 50-002-22966 Supplier Medchemexpress LLC Supplier No. HY1376625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5'-Phosphoguanylyl-(3',5')-guanosine (pGpG) is a nucleotide intermediate produced during enzymatic degradation of cyclic di-guanylate (c-di-GMP). It is supplied as a purified research reagent for biochemical and microbiology studies involving nucleotide signaling and enzymology.

  • Purity 95.8% suitable for analytical and biochemical assays.
  • Molecular weight 708.43 g/mol.
  • Chemical formula C20H26N10O15P2.
  • Available in milligram scales (5 mg, 10 mg, 25 mg, 50 mg, and larger sizes).
  • Intended for biochemical assays and studies of nucleotide signaling and c-di-GMP metabolism.
  • Provided with characterization data to support research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.