Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Phosphorylribose 1-pyrophosphate sodium salt hydrate, MP Biomedicals™

Catalog No. ICN10098125 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10098125 25 mg
ICN10098180 100 mg
ICN10098110 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10098125 Supplier MP Biomedicals, Inc Supplier No. 0210098125
Add to Cart
Add to Cart

Chemical Identifiers

CAS 108321-05-7
Molecular Formula C5H13NaO14P3
Molecular Weight (g/mol) 413.056
InChI Key CFIMWVBJOCHTCA-UHFFFAOYSA-N
Synonym alpha-d-ribosyldiphosphate5-phosphatepentasodiumsalt
PubChem CID 133611921
IUPAC Name [3,4-dihydroxy-5-(phosphonooxymethyl)oxolan-2-yl] phosphono hydrogen phosphate;sodium
SMILES C(C1C(C(C(O1)OP(=O)(O)OP(=O)(O)O)O)O)OP(=O)(O)O.[Na]

Specifications

Color White
Assay Percent Range 0.75
Quantity 25 mg
Formula Weight 390.1
Physical Form Powder
Chemical Name or Material 5-Phosphorylribose 1-pyrophosphate sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.