Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-(Piperazin-1-yl)isoquinoline hydrochloride | 209733-17-5 | 99.6% | 249.74 | 500 MG
SDP

Catalog No. 5000400833
Encompass_Preferred

5-(Piperazin-1-yl)isoquinoline hydrochloride is a chemical compound from MedChemExpress, identified as a drug intermediate. This product is characterized by its yellow to orange solid appearance.

  • Consistent with structure based on 1H NMR Spectrum and LCMS
  • Purity of 99.6% as determined by LCMS
  • Molecular formula: C13H16CIN3
  • Molecular weight: 249.74
  • Store at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture

Catalog No. 50-004-00833 Supplier Medchemexpress LLC Supplier No. HY23703A500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-(Piperazin-1-yl)isoquinoline hydrochloride is a chemical compound from MedChemExpress, identified as a drug intermediate. This product is characterized by its yellow to orange solid appearance.

  • Consistent with structure based on 1H NMR Spectrum and LCMS
  • Purity of 99.6% as determined by LCMS
  • Molecular formula: C13H16CIN3
  • Molecular weight: 249.74
  • Store at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.