Learn More
Cole-Parmer Cole-Parmer Five-Position Stirring Hot Plate, 120 VAC
Catalog No. NC0474659 Shop All Cole-Parmer Products
Encompass_Preferred
These multiposition stirring hot plates offer more versatility than single-position units. Stirring hot plates are available with five or nine stirring positions. Each stirring position is individually controlled so you can perform different mixing applications simultaneously. Use stirring hot plates to heat only, stir only, or heat and stir simultaneously. The corrosion-resistant ceramic tops heat quickly and never warp or buckle. The temperature control has OFF, HI, and LOW detents/settings, with 270 degrees of infinite control between the low and high detents.
Catalog No. NC0474659 Supplier Cole-Parmer Supplier No. 5145071
These multiposition stirring hot plates offer more versatility than single-position units. Stirring hot plates are available with five or nine stirring positions. Each stirring position is individually controlled so you can perform different mixing applications simultaneously. Use stirring hot plates to heat only, stir only, or heat and stir simultaneously. The corrosion-resistant ceramic tops heat quickly and never warp or buckle. The temperature control has OFF, HI, and LOW detents/settings, with 270 degrees of infinite control between the low and high detents.