Learn More
BIOCHEK USA CORP 5-plate poultry ELISA kit
Supplier:  BIOCHEK USA CORP CK124 ILT
Encompass
5-plate poultry ELISA kitTo detect antibodies to ILT virus in chickensPlates per kit: 5Maximum sample size: 460Run at room temperatureIncubation times: 60-60-30Read at 405 NM1:500 dilution
Catalog No. NC2744518