Learn More
Chemglass Life Sciences 5-Port Adapter, G 3/4 Thread, 30° Angled 1/8" Ports, Stainless Steel
SDP
Catalog No. 502733320
Encompass
The 5-Port inlet assembly fits into G 3/4 head plate port.The assembly incorporates five separate pipes into one assembly for efficient use of space.Each port can be used individually for reagent addition.G 3/4 threaded adapter has (5) 30 degrees Angled 1/8 (3.2mm) tubing ports for connection to flexible tubing.Inside the reactor, the 1/8 tubes are 1/8 long.
Catalog No. 50-273-3320 Supplier Chemglass Life Sciences Supplier No. CLS1380501
The 5-Port inlet assembly fits into G 3/4 head plate port.The assembly incorporates five separate pipes into one assembly for efficient use of space.Each port can be used individually for reagent addition.G 3/4 threaded adapter has (5) 30 degrees Angled 1/8 (3.2mm) tubing ports for connection to flexible tubing.Inside the reactor, the 1/8 tubes are 1/8 long.