Learn More
Chemglass Life Sciences 5-Port Adapter, Vertical Ports, G3/4 Thread, 1/8" Ports, Electro Polished Ra20, 316L Stainless Steel
SDP
Catalog No. 5000083145
Encompass
The 5-Port inlet assembly fits into G 3/4 head plate port.The assembly incorporates five separate pipes into one assembly for efficient use of space.Each port can be used individually for reagent addition.G 3/4 threaded adapter has (5) 1/8 (3. 2mm) tubing ports for connection to flexible tubing.Inside the reactor, the 1/8 tubes are 1/8 long.
Catalog No. 50-000-83145 Supplier Chemglass Life Sciences Supplier No. CLS1380500
The 5-Port inlet assembly fits into G 3/4 head plate port.The assembly incorporates five separate pipes into one assembly for efficient use of space.Each port can be used individually for reagent addition.G 3/4 threaded adapter has (5) 1/8 (3. 2mm) tubing ports for connection to flexible tubing.Inside the reactor, the 1/8 tubes are 1/8 long.