Learn More
Chemglass Life Sciences 5-Port Airfree Schlenk Vacuum Manifold, Single Bank, 465mm OAL
SDP
Catalog No. AF006103
Encompass_Preferred
5-Port Airfree Schlenk Vacuum Manifold, Single Bank, 465mm OAL. Single bank vacuum manifold with 0-8mm Chem-Vac™ Chem-Cap valves mounted on a 45 angle to the user permitting easy operation inside of a fume hood. Ports are spaced 75mm center to center. All connection ports are #15 o-ring connections that incorporate a greasefree o-ring seal. Manifold is modular in design allowing the user tremendous flexibility in performing experiments. Adapters from #15 o-ring joints to standard taper components, serrated hose connections or 3/8" Cajon fittings are available to allow the ports to be used with existing glassware. Replacement Chem-Vac™ Chem-Cap plug is listed below.
Catalog No. AF006103 Supplier Chemglass Life Sciences Supplier No. AF006103
5-Port Airfree Schlenk Vacuum Manifold, Single Bank, 465mm OAL. Single bank vacuum manifold with 0-8mm Chem-Vac™ Chem-Cap valves mounted on a 45 angle to the user permitting easy operation inside of a fume hood. Ports are spaced 75mm center to center. All connection ports are #15 o-ring connections that incorporate a greasefree o-ring seal. Manifold is modular in design allowing the user tremendous flexibility in performing experiments. Adapters from #15 o-ring joints to standard taper components, serrated hose connections or 3/8" Cajon fittings are available to allow the ports to be used with existing glassware. Replacement Chem-Vac™ Chem-Cap plug is listed below.