Learn More
Chemglass Life Sciences 5-Port Vacuum Manifold, Left Handed Trap Assembly, Double Bank, Airfree, Schlenk
SDP
Supplier:  Chemglass Life Sciences AF045002
Encompass_Preferred
5-Port Vacuum Manifold, Left Handed Trap Assembly, Double Bank, Airfree, Schlenk, Complete system consists of one 5-port vacuum/inert manifold, one pre-trap, one main trap assembly, one main trap body, two CG-150-05 #28 S.S.pinch clamps, one CG-150-06 #35 S.S.pinch clamp, one CG-150-21 #75 S.S.pinch clamp, two CG-110-01 1/2" S.S.springs, one 14/20 outer stopper with hooks, and one CG-3014-05 35/25 socket stopper. System is available in right or left handed configurations. Overall dimensions are 40" W x 24" H.
Catalog No. AF045002
Add to Cart
Request a Quote