Learn More
Bellco Glass, Inc. 5 Position Digital CPT Stir Complete, 115V
SDP
Catalog No. 501946234
Encompass_Preferred
Bell-ennium Magnetic Stirrers are designed for both cell cultures in spinner flasks as well as for general laboratory procedures requiring the use of magnetic stirring equipment. Bell-ennium stirrers offer the laboratory researcher a wide range of capabilities and the latest innovations in stirring technology. Bell-ennium stirrers feature brushless maintenance free DC motors with drive assemblies designed for quiet operation. Stainless steel covers are stamped to show the vessel positions for accurate placement and optimum performance. The Bell-ennium single position low profile chassis measure 10 x 11 x 3 3/8” and are designed to fit side-by-side in Bellco incubators. Units are powered by standard 24V in-line power supplies and are CE compliant.. MADE IN THE USA
Catalog No. 50-194-6234 Supplier Bellco Glass, Inc. Supplier No. 7785D5005
Bell-ennium Magnetic Stirrers are designed for both cell cultures in spinner flasks as well as for general laboratory procedures requiring the use of magnetic stirring equipment. Bell-ennium stirrers offer the laboratory researcher a wide range of capabilities and the latest innovations in stirring technology. Bell-ennium stirrers feature brushless maintenance free DC motors with drive assemblies designed for quiet operation. Stainless steel covers are stamped to show the vessel positions for accurate placement and optimum performance. The Bell-ennium single position low profile chassis measure 10 x 11 x 3 3/8” and are designed to fit side-by-side in Bellco incubators. Units are powered by standard 24V in-line power supplies and are CE compliant.. MADE IN THE USA