Learn More
Medchemexpress LLC HY-108213A 100mg Medchemexpress, Inosine 5'-monophosphate (disodium) salt (hydrate) CAS:352195-40-5 Purity:>98%
SDP
Catalog No. 501968511
Encompass_Preferred
Medchemexpress, HY-109564 10mg 5α-Pregnane-3β,6α-diol-20-one CAS:21853-11-2 5α-Pregnane-3β,6α-diol-20-one is a mitogenic metabolite of progesterone, and it can be produced in starved androgen-responsive prostate cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8511 Supplier Medchemexpress LLC Supplier No. HY108213A100MG
Medchemexpress, HY-109564 10mg 5α-Pregnane-3β,6α-diol-20-one CAS:21853-11-2 5α-Pregnane-3β,6α-diol-20-one is a mitogenic metabolite of progesterone, and it can be produced in starved androgen-responsive prostate cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.