Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-[(prop-2-yn-1-yl)amino]-2,3-dihydro-1H-isoindole-1,3-dione | 2357110-24-6 | MFCD34563706 | 98.0% | 311.29 g/mol | C16H13N3O4 | 50 MG
SDP

Catalog No. 5000284650
Encompass_Preferred

Thalidomide-5-NH-C-alkyne is an E3 ligase ligand-linker conjugate that contains a terminal alkyne for bioorthogonal conjugation. It is provided as a small-quantity research reagent intended for PROTAC development and target protein conjugation. Reported properties include a molecular formula of C16H13N3O4, a molecular weight of 311.29 g/mol, and a reported purity of 98.0%.

  • Contains an E3 ligase ligand with a terminal alkyne for conjugation.
  • Designed for PROTAC development and target protein degradation studies.
  • Reported purity 98.0% suitable for research applications.
  • Provided as a small-quantity reagent (50 mg) for synthetic chemistry workflows.
  • Molecular formula C16H13N3O4; molecular weight 311.29 g/mol.
  • Store and handle according to supplier safety data sheet recommendations.

Catalog No. 50-002-84650 Supplier Medchemexpress LLC Supplier No. HYW67432450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-5-NH-C-alkyne is an E3 ligase ligand-linker conjugate that contains a terminal alkyne for bioorthogonal conjugation. It is provided as a small-quantity research reagent intended for PROTAC development and target protein conjugation. Reported properties include a molecular formula of C16H13N3O4, a molecular weight of 311.29 g/mol, and a reported purity of 98.0%.

  • Contains an E3 ligase ligand with a terminal alkyne for conjugation.
  • Designed for PROTAC development and target protein degradation studies.
  • Reported purity 98.0% suitable for research applications.
  • Provided as a small-quantity reagent (50 mg) for synthetic chemistry workflows.
  • Molecular formula C16H13N3O4; molecular weight 311.29 g/mol.
  • Store and handle according to supplier safety data sheet recommendations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.