Learn More
Medchemexpress LLC 5-propargylamino-dCTP | 115899-39-3 | MFCD28010326 | 99.5% | 520.22 g/mol | C12H19N4O13P3 | 5 MG
SDP
Catalog No. 5000262820
Encompass_Preferred
5-Propargylamino-dCTP is an alkyne-modified deoxycytidine triphosphate designed for incorporation into DNA and subsequent conjugation using click chemistry. It is intended for research applications requiring sensitive nucleic acid labeling and high chemical purity.
  • Alkyne-modified nucleotide for bioorthogonal conjugation.
  • Enables click chemistry labeling of incorporated nucleotides.
  • High purity suitable for molecular biology assays.
  • Water soluble at high concentration for convenient formulation.
  • Stable when stored frozen; follow sealed, moisture-free storage.
Catalog No. 50-002-62820 Supplier Medchemexpress LLC Supplier No. HY1321425MG
Add to Cart
Add to Cart
5-Propargylamino-dCTP is an alkyne-modified deoxycytidine triphosphate designed for incorporation into DNA and subsequent conjugation using click chemistry. It is intended for research applications requiring sensitive nucleic acid labeling and high chemical purity.
  • Alkyne-modified nucleotide for bioorthogonal conjugation.
  • Enables click chemistry labeling of incorporated nucleotides.
  • High purity suitable for molecular biology assays.
  • Water soluble at high concentration for convenient formulation.
  • Stable when stored frozen; follow sealed, moisture-free storage.