Learn More
Medchemexpress LLC 5-propargylamino-dCTP | 115899-39-3 | MFCD28010326 | 99.5% | C12H19N4O13P3 | 10MG
SDP
Catalog No. 5000260598
Encompass_Preferred
5-Propargylamino-dCTP is an alkyne-functionalized deoxycytidine triphosphate designed for enzymatic incorporation into DNA and for subsequent covalent labeling via click chemistry. The terminal alkyne enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) for attachment of probes or tags used in detection, sequencing, and imaging workflows.
  • Alkyne handle compatible with copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Designed for enzymatic incorporation into DNA as a dCTP analog.
  • Enables post-synthetic conjugation of fluorescent probes or affinity tags.
  • High purity for reliable biochemical performance.
  • Soluble in water to facilitate aqueous labeling reactions.
  • Suitable for nucleic acid labeling, sequence analysis, and probe development.
Catalog No. 50-002-60598 Supplier Medchemexpress LLC Supplier No. HY13214210MG
5-Propargylamino-dCTP is an alkyne-functionalized deoxycytidine triphosphate designed for enzymatic incorporation into DNA and for subsequent covalent labeling via click chemistry. The terminal alkyne enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) for attachment of probes or tags used in detection, sequencing, and imaging workflows.
  • Alkyne handle compatible with copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Designed for enzymatic incorporation into DNA as a dCTP analog.
  • Enables post-synthetic conjugation of fluorescent probes or affinity tags.
  • High purity for reliable biochemical performance.
  • Soluble in water to facilitate aqueous labeling reactions.
  • Suitable for nucleic acid labeling, sequence analysis, and probe development.