Learn More
Apexbio Technology LLC 5-Propargylamino-dCTP 50ul (100 mM)
SDP
Catalog No. 5000088040
Encompass_Preferred
5-Propargylamino-dCTP is a modified nucleoside triphosphate analog featuring a cytosine base with a propargylamino substituent introducing an alkyne functional group 5-Propargylamino-dCTP is incorporated into DNA strands mediated by DNA polymerases during active DNA synthesis This modification enables covalent conjugation to azide-bearing probes via copper-catalyzed azide-alkyne cycloaddition (CuAAC) Based on these pharmacological properties 5-Propargylamino-dCTP holds research potential in the detection and analysis of cellular proliferation DNA replication dynamics nucleic acid repair subcellular localization studies fluorescence labeling and molecular imaging-based nucleic acid research methodologies
Catalog No. 50-000-88040 Supplier Apexbio Technology LLC Supplier No. B81140.05
5-Propargylamino-dCTP is a modified nucleoside triphosphate analog featuring a cytosine base with a propargylamino substituent introducing an alkyne functional group 5-Propargylamino-dCTP is incorporated into DNA strands mediated by DNA polymerases during active DNA synthesis This modification enables covalent conjugation to azide-bearing probes via copper-catalyzed azide-alkyne cycloaddition (CuAAC) Based on these pharmacological properties 5-Propargylamino-dCTP holds research potential in the detection and analysis of cellular proliferation DNA replication dynamics nucleic acid repair subcellular localization studies fluorescence labeling and molecular imaging-based nucleic acid research methodologies