Learn More
Medchemexpress LLC 5-Propargyltryptamide formic | 2906227-87-8 | 99.9% | 260.29 g/mol | C14H16N2O3 | 10 MG
SDP
Catalog No. 5000287847
Encompass_Preferred
5-PT formic (5-Propargyltryptamide formic) is a serotonin-derived chemical probe containing a propargyl (alkyne) moiety for copper-catalyzed azide-alkyne cycloaddition (click chemistry). It is used to label and study protein serotonylation in biochemical and cell-based assays.
  • Enables copper-catalyzed click chemistry labeling of serotonylated proteins.
  • High chemical purity (99.91%).
  • Solid, white to off-white physical form for easy handling and storage.
  • Suitable for biochemical and cell-based assay applications.
  • Available in small-scale quantities (for example, 10 mg) for exploratory studies.
Catalog No. 50-002-87847 Supplier Medchemexpress LLC Supplier No. HY156562A10MG
5-PT formic (5-Propargyltryptamide formic) is a serotonin-derived chemical probe containing a propargyl (alkyne) moiety for copper-catalyzed azide-alkyne cycloaddition (click chemistry). It is used to label and study protein serotonylation in biochemical and cell-based assays.
  • Enables copper-catalyzed click chemistry labeling of serotonylated proteins.
  • High chemical purity (99.91%).
  • Solid, white to off-white physical form for easy handling and storage.
  • Suitable for biochemical and cell-based assay applications.
  • Available in small-scale quantities (for example, 10 mg) for exploratory studies.