Learn More
Apexbio Technology LLC 5-Propynyl-dUTP 50ul (100 mM)
SDP
Catalog No. 5000063674
Encompass_Preferred
5-Propynyl-dUTP is a nucleotide analog modified by the addition of a propynyl functional group at the 5-position of the pyrimidine ring This chemical modification confers altered interactions with DNA polymerases during enzymatic DNA synthesis Due to its structural alteration 5-Propynyl-dUTP can be incorporated into DNA strands during replication or in vitro polymerase reactions thereby allowing targeted modulation of DNA polymerase activity and DNA extension dynamics Applications in molecular biology and genetic research include investigating DNA replication processes examining DNA repair pathways detecting mutation events and probing molecular interactions in diverse cellular contexts
Catalog No. 50-000-63674 Supplier Apexbio Technology LLC Supplier No. B80880.05
5-Propynyl-dUTP is a nucleotide analog modified by the addition of a propynyl functional group at the 5-position of the pyrimidine ring This chemical modification confers altered interactions with DNA polymerases during enzymatic DNA synthesis Due to its structural alteration 5-Propynyl-dUTP can be incorporated into DNA strands during replication or in vitro polymerase reactions thereby allowing targeted modulation of DNA polymerase activity and DNA extension dynamics Applications in molecular biology and genetic research include investigating DNA replication processes examining DNA repair pathways detecting mutation events and probing molecular interactions in diverse cellular contexts