Learn More
Medchemexpress LLC 5-Pyrimidineacetic acid, 4,6-bis(dimethylamino)-2-[[4-[[4-(trifluoromethyl)benzoyl]amino]phenyl]methyl]- | 1093108-50-9 | 99.4% | 501.50 | 25 MG
SDP
Catalog No. 5000438367
Encompass_Preferred
BI-671800 is a highly specific and potent antagonist of chemoattractant receptor-homologous molecule on Th2 cells (DP2/CRTH2), with IC50 values of 4.5 nM and 3.7 nM for PGD2 binding to CRTH2 in hCRTH2 and mCRTH2 transfected cells, respectively. It has potential for the treatment of poorly controlled asthma.
  • Highly specific and potent antagonist of chemoattractant receptor-homologous molecule on Th2 cells (DP2/CRTH2).
  • IC50 values of 4.5 nM and 3.7 nM for PGD2 binding to CRTH2 in hCRTH2 and mCRTH2 transfected cells, respectively.
  • Potential for the treatment of poorly controlled asthma.
  • Solid appearance, white to off-white color.
Catalog No. 50-004-38367 Supplier Medchemexpress LLC Supplier No. 25MGHY114141
BI-671800 is a highly specific and potent antagonist of chemoattractant receptor-homologous molecule on Th2 cells (DP2/CRTH2), with IC50 values of 4.5 nM and 3.7 nM for PGD2 binding to CRTH2 in hCRTH2 and mCRTH2 transfected cells, respectively. It has potential for the treatment of poorly controlled asthma.
  • Highly specific and potent antagonist of chemoattractant receptor-homologous molecule on Th2 cells (DP2/CRTH2).
  • IC50 values of 4.5 nM and 3.7 nM for PGD2 binding to CRTH2 in hCRTH2 and mCRTH2 transfected cells, respectively.
  • Potential for the treatment of poorly controlled asthma.
  • Solid appearance, white to off-white color.