Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Pyrimidinecarbonitrile, 4-[4-cyano-2-(methylsulfonyl)phenyl]-1,2,3,4-tetrahydro-3,6-dimethyl- | 2446175-39-7 | 99.1% | 474.46 | 10 MM 1 ML
SDP

Catalog No. 5000401803
Encompass_Preferred

(R)-BAY-85-8501 is the less active enantiomer of BAY-85-8501. BAY-85-8501 is a selective and potent inhibitor of Human Neutrophil Elastase (HNE), with an IC50 of 65 pM. This product is for research use only.

  • High purity: 99.09%.
  • Appearance: White to off-white solid.
  • Molecular formula: C22H17F3N4O3S.
  • Soluble in DMSO (100 mg/mL).
  • Long-term storage recommended at -20°C for powder or -80°C for solutions.

Catalog No. 50-004-01803 Supplier Medchemexpress LLC Supplier No. HY1990810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-BAY-85-8501 is the less active enantiomer of BAY-85-8501. BAY-85-8501 is a selective and potent inhibitor of Human Neutrophil Elastase (HNE), with an IC50 of 65 pM. This product is for research use only.

  • High purity: 99.09%.
  • Appearance: White to off-white solid.
  • Molecular formula: C22H17F3N4O3S.
  • Soluble in DMSO (100 mg/mL).
  • Long-term storage recommended at -20°C for powder or -80°C for solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.