Learn More
Medchemexpress LLC 5-Pyrimidinecarboxamide, 2-[4-[[[2-(2,5-dimethyl-3-furanyl)-6,7-dimethoxy-4-quinolinyl]amino]methyl | 2256079-39-5 | 98.4% | 532.59 | 25 MG
SDP
Catalog No. 5000391895
Encompass_Preferred
CM-1758 is a histone deacetylase (HDAC) inhibitor. It inhibits tumor growth in vivo and induces acetylation of non-histone proteins in acute myeloid leukemia cells.
  • Histone deacetylase (HDAC) inhibitor
  • Inhibits tumor growth in vivo
  • Induces acetylation of non-histone proteins in acute myeloid leukemia cells
  • For research use only
Catalog No. 50-003-91895 Supplier Medchemexpress LLC Supplier No. 25MGHY161149
CM-1758 is a histone deacetylase (HDAC) inhibitor. It inhibits tumor growth in vivo and induces acetylation of non-histone proteins in acute myeloid leukemia cells.
  • Histone deacetylase (HDAC) inhibitor
  • Inhibits tumor growth in vivo
  • Induces acetylation of non-histone proteins in acute myeloid leukemia cells
  • For research use only