Learn More
Medchemexpress LLC 5-Pyrimidinecarboxamide, N-(2-chloro-6-methylphenyl)-2-[[4-(4-methyl-1-piperazinyl)phenyl]amino]-... | 2556836-50-9 | 98.6% | 597.11 | 100 MG
SDP
Catalog No. 5000350080
Encompass_Preferred
DGY-06-116 is an irreversible covalent and selective Src inhibitor with an IC50 of 3 nM. It demonstrates potent antiproliferative effects in non-small cell lung cancer (NSCLC) and triple-negative breast cancer (TNBC) cell lines, effectively inhibiting Src kinase activity and signaling.
  • Irreversible covalent and selective Src inhibitor
  • Potent antiproliferative effects in various cancer cell lines
  • Induces potent Src binding and inhibition of Src signaling
  • Effective in vivo inhibition of Src with a short half-life
Catalog No. 50-003-50080 Supplier Medchemexpress LLC Supplier No. HY136605100MG
DGY-06-116 is an irreversible covalent and selective Src inhibitor with an IC50 of 3 nM. It demonstrates potent antiproliferative effects in non-small cell lung cancer (NSCLC) and triple-negative breast cancer (TNBC) cell lines, effectively inhibiting Src kinase activity and signaling.
  • Irreversible covalent and selective Src inhibitor
  • Potent antiproliferative effects in various cancer cell lines
  • Induces potent Src binding and inhibition of Src signaling
  • Effective in vivo inhibition of Src with a short half-life