Learn More
Medchemexpress LLC 5-pyrimidinecarboxamide, N-(2-chloro-6-methylphenyl)-2-[[4-(4-methyl-1-piperazinyl)phenyl]amino]-4-[[2-[(1-oxo-2-propen-1-yl)amino]phenyl]amino]- | 2556836-50-9 | 98.6% | 597.11 | 25 MG
SDP
Catalog No. 5000350619
Encompass_Preferred
DGY-06-116 is an irreversible covalent, selective Src inhibitor with an IC50 of 3 nM. It also inhibits FGFR1 with an IC50 of 8340 nM. This product is for research use only.
  • Selective Src inhibitor
  • Inhibits FGFR1
  • White to off-white solid appearance
Catalog No. 50-003-50619 Supplier Medchemexpress LLC Supplier No. 25MGHY136605
DGY-06-116 is an irreversible covalent, selective Src inhibitor with an IC50 of 3 nM. It also inhibits FGFR1 with an IC50 of 8340 nM. This product is for research use only.
  • Selective Src inhibitor
  • Inhibits FGFR1
  • White to off-white solid appearance