Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Pyrimidinylboronic acid, 97%, Thermo Scientific™

Catalog No. CC07412DA
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC07412DA 1 g Amber Glass Bottle
CC07412CB 250 mg Amber Glass Bottle
CC07412EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC07412DA Supplier Thermo Scientific Chemicals Supplier No. CC07412DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 109299-78-7
Molecular Formula C4H5BN2O2
Molecular Weight (g/mol) 123.91
MDL Number MFCD03002366
InChI Key HZFPPBMKGYINDF-UHFFFAOYSA-N
Synonym 5-pyrimidinylboronic acid, pyrimidine-5-boronic acid, 5-pyrimidineboronic acid, 5-pyrimidylboronic acid, pyrimidin-5-yl boronic acid, 5-pyrimidine boronic acid, pyrimidinyl-5-boronic acid, pyrimidine-5-ylboronic acid, pyrimidin-5-yl-5-boronic acid, pyrimidin-5-boronic acid
PubChem CID 2795193
SMILES OB(O)C1=CN=CN=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name pyrimidin-5-ylboronic acid
Formula Weight 123.91
Percent Purity 97%
Chemical Name or Material 5-Pyrimidinylboronic acid
Hazard Category Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.