Learn More
Medchemexpress LLC 5-Pyrimidylboronic acid | 109299-78-7 | 99.9% | C4H5BN2O2 | 10 G
SDP
Catalog No. 5000434004
Encompass_Preferred
5-Pyrimidylboronic acid is a biochemical reagent suitable for life science related research, where it can be utilized as a biological material or organic compound.
  • Functions as a biochemical reagent.
  • Can be employed as a biological material.
  • Can be used as an organic compound for life science related research.
  • Applicable in analogous one-pot copper-catalyzed DBAD coupling and condensation reactions.
  • Facilitates Suzuki-Miyaura coupling, allowing for the coupling of boronic acids with carbon halides and surrogates.
Catalog No. 50-004-34004 Supplier Medchemexpress LLC Supplier No. HY5922610G
5-Pyrimidylboronic acid is a biochemical reagent suitable for life science related research, where it can be utilized as a biological material or organic compound.
  • Functions as a biochemical reagent.
  • Can be employed as a biological material.
  • Can be used as an organic compound for life science related research.
  • Applicable in analogous one-pot copper-catalyzed DBAD coupling and condensation reactions.
  • Facilitates Suzuki-Miyaura coupling, allowing for the coupling of boronic acids with carbon halides and surrogates.