Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Pyrrolidin-1-ylpyridine-2-carboxylic acid, 95%, Thermo Scientific™

Catalog No. CC52901DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC52901DA 1 g Amber Glass Bottle
CC52901CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC52901DA Supplier Thermo Scientific Chemicals Supplier No. CC52901DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 950603-19-7
Molecular Formula C10H12N2O2
Molecular Weight (g/mol) 192.22
MDL Number MFCD09879903
InChI Key ACLFTMQROMOTLA-UHFFFAOYSA-N
Synonym 5-pyrrolidin-1-yl picolinic acid, 5-pyrrolidin-1-yl pyridine-2-carboxylic acid, pubchem19497, 5-pyrrolidinylpyridine-2-carboxylic acid, 2-pyridinecarboxylicacid,5-1-pyrrolidinyl, 5-pyrrolidin-1-yl-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 5-1-pyrrolidinyl
PubChem CID 24229530
SMILES OC(=O)C1=NC=C(C=C1)N1CCCC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 5-(pyrrolidin-1-yl)pyridine-2-carboxylic acid
Formula Weight 192.22
Percent Purity 95%
Chemical Name or Material 5-Pyrrolidin-1-ylpyridine-2-carboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.