Learn More
STERICYCLE 5 Quart Covidien RCRA Container
Catalog No. NC3715896
Encompass
5 Quart Covidien RCRA Container Lid: Black. Dimensions: 6"x11"x13.25". Designed for the safe collection, containment and segregation of hazardous pharmaceutical waste to comply with the Resource Conservation and Recovery Act (RCRA)
Catalog No. NC3715896 Supplier STERICYCLE Supplier No. 8605RC
5 Quart Covidien RCRA Container Lid: Black. Dimensions: 6"x11"x13.25". Designed for the safe collection, containment and segregation of hazardous pharmaceutical waste to comply with the Resource Conservation and Recovery Act (RCRA)