Learn More
Medchemexpress LLC 5-Quinolinylboronic acid | 355386-94-6 | 172.98 | 1 G
SDP
Catalog No. 5000445599
Encompass_Preferred
5-Quinolinylboronic acid, also known as Quinolin-5-ylboronic, is a biochemical reagent. It can be utilized as a biological material or organic compound for research within the life sciences.
  • Used as a biochemical reagent
  • Suitable as a biological material or organic compound
  • Appears as a solid with color ranging from white to light yellow
  • Soluble in DMSO at 50 mg/mL
  • For research use only
Catalog No. 50-004-45599 Supplier Medchemexpress LLC Supplier No. HYW0030171G
5-Quinolinylboronic acid, also known as Quinolin-5-ylboronic, is a biochemical reagent. It can be utilized as a biological material or organic compound for research within the life sciences.
  • Used as a biochemical reagent
  • Suitable as a biological material or organic compound
  • Appears as a solid with color ranging from white to light yellow
  • Soluble in DMSO at 50 mg/mL
  • For research use only