Learn More
Apexbio Technology LLC 5-R-Rivaroxaban, 10mg. Cas: 865479-71-6 MFCD: MFCD17171369. Factor Xa (FXa) inhibitor
SDP
Supplier:  Apexbio Technology LLC A312610
Encompass_Preferred
5-R-Rivaroxaban is a selective inhibitor of human Factor Xa with IC50 value of 0.7 nmol/L [1]. Factor Xa is a serine endopeptidase enzyme and plays an important role in the convergence point of the intrinsic and extrinsic pathways in blood coagulation system [2]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4786