Learn More
LIFECANVAS TECHNOLOGIES INC 5 ROUNDS STAINING BUFFER KIT
SDP
Supplier:  LIFECANVAS TECHNOLOGIES INC BF1L01
Encompass
Reagents:5 Rounds Staining Buffer Kit - 5 x Primary Buffer - 5 x Wash Buffer - 500 mL Primary Sample Buffer - 500 mL Sample Wash Buffer- Special Formulation for Kidney Samples
Catalog No. NC1908112