Learn More
AdipoGen 5-ROX
SDP
Supplier:  AdipoGen CDXC0004M020
Encompass_Preferred
Chemical. CAS 216699-35-3. Formula C33H30N2O5. MW 534.6. Synthetic. 5-ROX is the purified isomer of 56-ROX. It is preferred for some complicated biological applications where reproducibility is really critical. The minor positional difference between 5-ROX and 6-ROX might significantly affect some biological properties of the underlying conjugates. Used for oligonucleotide labeling and automated DNA sequencing applications.
Catalog No. 50-256-1566