Learn More
AdipoGen 5-ROX N-succinimidyl ester
SDP
Supplier:  AdipoGen CDXC0019M005
Encompass_Preferred
Chemical. CAS 209734-74-7. Formula C37H33N3O7. MW 631.67. 5-ROX N-succinimidyl ester 5-ROX-SE is an amine-reactive long wavelength rhodamine dye. Derivatives made with the amine-reactive carboxy-X-rhodamine ROX, succinimidyl ester are widely used for oligonucleotide labeling and automated DNA sequencing applications. Conjugates of this dye have longer-wavelength spectra than the spectra of Lissamine rhodamine B conjugates, but somewhat shorter-wavelength spectra than those of Texas Red conjugates. 5-ROX-SE derivatized compounds such as proteins, nucleic acids and drugs may be analyzed by fluorescence resonance energy transfer FRET and fluorescence quenching applications. 5-ROX-SE is a labeling reagent for preparation of charge-modified dye-labeled ddNTPs to "direct-load" DNA sequencing.
Catalog No. 50-256-1544