Learn More
Aat Bioquest 5-ROX, SE [5-Carboxy-X-rhodamine, succinimidyl ester] *CAS 209734-74-7*
SDP
Supplier:  Aat Bioquest 389
Encompass_Preferred
Single isomers of ROX are increasingly preferred for labeling peptides and nucleotides because they give better resolution in HPLC purifications that are often required in bioconjugations.
Catalog No. 50-195-2871