Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences 0.5% Ruthenium Tetroxide Aqueous 10 ML
SDP

Catalog No. 5028412
Encompass_Preferred

RuO₄ crystal form F.W. 165.70, crystal form
Melting point: 25.4°C
Boiling point: 40°C

Ruthenium tetroxide is very similar to Osmium tetroxide, and is used as a fixation/staining agent for electron microscopy. Ruthenium tetroxide offers excellent staining of saturated and unsaturated polymer materials as well as stabilizing thin sections under the bombardment of the electron beam. NOTE: Penetration of ruthenium tetroxide into the tissue is poor.

Catalog No. 50-284-12 Supplier Electron Microscopy Sciences Supplier No. 2070005
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RuO₄ crystal form F.W. 165.70, crystal form
Melting point: 25.4°C
Boiling point: 40°C

Ruthenium tetroxide is very similar to Osmium tetroxide, and is used as a fixation/staining agent for electron microscopy. Ruthenium tetroxide offers excellent staining of saturated and unsaturated polymer materials as well as stabilizing thin sections under the bombardment of the electron beam. NOTE: Penetration of ruthenium tetroxide into the tissue is poor.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.