Learn More
Enzo Life Sciences 5(S),12(R)-Dihydroxyeicosa-6E,8E,10E,14Z-tetraenoic acid (50µg). CAS: 71652-82-9
SDP
Supplier:  Enzo Life Sciences BMLDTR5120050
Encompass_Preferred
Biosynthesis via nonenzymatic and enzymatic hydrolysis of LTA4 and by myeloperoxidase-dependent metabolism of cysteinyl leukotrienes. Neutrophil chemotaxin in the guinea pig dermis. Metabolite of LTB4 in rat kidney produced enzymatically by a novel isomerase reaction. Alternative name: 5(S),12(R)-DiHETE. Formulation: Oil dissolved in ethanol. Purity: ≥99% (HPLC). Long Term Storage: -80°C.
Catalog No. 50-201-0520