Learn More
Cayman Chemical 5 S 15 S-DIHETE 100ug
SDP
Catalog No. 502913704
Encompass_Preferred
A dihydroxy fatty acid synthesized by 15-LO from 5(S)-HETE; chemotactic for eosinophils (ED50 = 0.3 µM)
Catalog No. 50-291-3704 Supplier Cayman Chemical Supplier No. 35280100 UG
A dihydroxy fatty acid synthesized by 15-LO from 5(S)-HETE; chemotactic for eosinophils (ED50 = 0.3 µM)